missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-11 Antibody (RM0256-22C39), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11656G
This item is not returnable.
View return policy
Description
IL-11 Monoclonal specifically detects IL-11 in Mouse samples. It is validated for Western Blot.
Specifications
| IL-11 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IL11 | |
| Mouse recombinant IL-11 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0256-22C39 | |
| Western Blot | |
| Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11Oprelvekin, interleukin 11, interleukin-11 | |
| Rat | |
| Protein A or G purified | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| 3589 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction