missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 Antibody (1-1A4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50440AF700
This item is not returnable.
View return policy
Description
IL-12 Monoclonal specifically detects IL-12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IL-12 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IL12A | |
| Recombinant human IL-12 produced in Baculovirus. | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 3592 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 1-1A4 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Binds p40 subunit of IL-12 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction