missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12/IL-23 p40 Antibody (240) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26108
This item is not returnable.
View return policy
Description
IL-12/IL-23 p40 Monoclonal antibody specifically detects IL-12/IL-23 p40 in Mouse samples. It is validated for ELISA
Specifications
| IL-12/IL-23 p40 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 240 | |
| ELISA 1:5000-1:10000 | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
| Rabbit immunized with purified, recombinant Mouse IL-12 p40/IL12B (Uniprot#: P43432, Met1-Ser335) | |
| 100 μL | |
| Innate Immunity | |
| 3593 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction