Learn More
IL-12/IL-23 p40 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30956-1-AP-20UL
Description
The IL-12/IL-23 p40 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IL-12/IL-23 p40. This antibody recognizes human antigen. The IL-12/IL-23 p40 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IL-12/IL-23 p40 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| CLMF, CLMF p40, CLMF2, IL 12 subunit p40, IL 12B, IL-12/IL-23 p40, IL12B, IL-12B, IL-12B/p40, Interleukin 12 subunit beta, NKSF, NKSF2 | |
| 20 μL | |
| 3593 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |