Learn More
IL-12/IL-23 p40 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30956-1-AP-150UL
 View more versions of this product
Product Code. 18096865
  • € 410.00 / 150µL
Description
The IL-12/IL-23 p40 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IL-12/IL-23 p40. This antibody recognizes human antigen. The IL-12/IL-23 p40 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
IL-12/IL-23 p40
400 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
CLMF, CLMF p40, CLMF2, IL 12 subunit p40, IL 12B, IL-12/IL-23 p40, IL12B, IL-12B, IL-12B/p40, Interleukin 12 subunit beta, NKSF, NKSF2
150 μL
3593
Store at -20°C. Stable for one year after shipment.