Learn More
Invitrogen™ IL-12/IL-23 p40 (Total) Mouse Uncoated ELISA Kit
Uncoated ELISA Kit
€ 512.00 - € 738.00
Specifications
Accession Number | P43432,Q9EQ14 |
---|---|
Assay Range | 2-300 pg/mL |
Assay Sensitivity | 2 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15521057
|
Invitrogen™
88-7120-77 |
20 x 96 Tests |
€ 738.00
Pack of 20 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
15541057
|
Invitrogen™
88-7120-88 |
10 x 96 Tests |
€ 512.00
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
The Mouse IL-12/IL-23 p40 (Total) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-12/IL-23 p40 (Total). Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IL-12 is an interleukin produced by dendritic cells, neutrophils, macrophages and human lymphoblastoid cells as a result of antigenic stimulation. IL-12 is a heterodimeric cytokine encoded by IL-12A (p35) and IL-12B (p40). IL-23 is a heterodimeric cytokine that shares a subunit with IL-12 (IL-12B) along with an IL-23A (IL-23p19) subunit. Functionally, IL-12 increases the expression of vascular endothelial adhesion molecule-1 on the endothelium, directs leukocytes towards tumors and facilitates the ischemic-hemorrhagic necrosis of the tumor tissue. In comparison, IL-23 plays an important role in stimulating memory T-cells, and is required for the induction, expansion, maintenance and downstream effector functions of Th17 cells, which play a vital role in upregulating neutrophil chemokines and various pro-inflammatory cytokines.Specifications
P43432,Q9EQ14 | |
2 pg/mL | |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
IL-23,p19,Il-12b,Il-12p40,Il12p40,p40 | |
HRP | |
Serum, 100 μL; Supernatant, 100 μL | |
Interleukin-12, IL12, IL23 | |
1 hr. 45 min. |
2-300 pg/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
16160, 83430 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, ELISA/ELISPOT Coating Buffer Powder: This uncoated ELISA kit may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20)., Assay Diluent: 5X concentrated, Detection enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -44; -76; -86) | |
RUO | |
2°C to 8°C | |
Mouse | |
24 hr. 30 min. |