missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12/IL-35 p35 Antibody (116219), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1171D
This item is not returnable.
View return policy
Description
IL-12/IL-35 p35 Monoclonal antibody specifically detects IL-12/IL-35 p35 in Porcine samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| IL-12/IL-35 p35 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| 116219 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CLMF,IL12A,IL-12A,IL-12p35,Ll12a,NFSK,NKSF1,P35 | |
| Chinese hamster ovary cell line CHO-derived recombinant porcine IL-12 p35/p40 heterodimer, p40 Ile23-Asn324, p35 Arg26-Ser222, Accession # Q28938, Q29053 | |
| 0.5 mL | |
| Innate Immunity | |
| Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction