missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12/IL-35 p35 Antibody (116219) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1171JF525
This item is not returnable.
View return policy
Description
IL-12/IL-35 p35 Monoclonal antibody specifically detects IL-12/IL-35 p35 in Porcine samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| IL-12/IL-35 p35 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CLMF;IL12A;IL-12A;IL-12p35;Ll12a;NFSK;NKSF1;P35 | |
| Chinese hamster ovary cell line CHO-derived recombinant porcine IL-12 p35/p40 heterodimer, p40 Ile23-Asn324, p35 Arg26-Ser222, Accession # Q28938, Q29053 | |
| 0.5 mL | |
| Innate Immunity | |
| Pig | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 116219 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction