missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12/IL-35 p35 Antibody (27537), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1570CL1
This item is not returnable.
View return policy
Description
IL-12/IL-35 p35 Monoclonal antibody specifically detects IL-12/IL-35 p35 in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| IL-12/IL-35 p35 | |
| Monoclonal | |
| CoraFluor 1 | |
| CLMF;IL12A;IL-12A;IL-12p35;Ll12a;NFSK;NKSF1;P35 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12/IL-35 p35, Arg23-Ser219, Accession # P29459 | |
| 0.1 mL | |
| Innate Immunity | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 27537 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction