missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-12 p35 Monoclonal Antibody (C3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543743
This item is not returnable.
View return policy
Description
IL-12 p35 Monoclonal Antibody for Western Blot
Interleukin 12A is a novel signaling cytokine belonging to the Interleukin-6 protein superfamily. Interleukin-12 alpha chain precursor (IL-12A) is a 35-kD subunit of disulfide-linked heterodimer of cytokine IL-12 and has a broad array of biological activities. Reports suggest that IL-12A can act as a growth factor for activated T and NK cells and enhance the lytic activity of NK/lymphokine-activated Killer cells. IL-12A stimulates the production of IFN-gamma by resting PBMC as well as T-cell-independent induction of interferon (IFN)-gamma, and helps in the differentiation of both Th1 and Th2 cells. It is also involved in positive regulation of tyrosine phosphorylation of Stat4 protein. The signaling process of IL-12A in innate immunity requires nitric oxide synthase 2A. Along with EBI3, IL-12A forms a novel heterodimeric cytokine, designated as interleukin-35 (IL-35), that may be specifically produced by T reg cells and is required for maximal suppressive activity.
Specifications
| IL-12 p35 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P29459, P43431 | |
| IL12A | |
| Recombinant protein Interleukin 12A. The antigen corresponds to amino acid range 23-219 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C3 | |
| Unconjugated | |
| IL12A | |
| CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 16159, 3592 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction