missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 R beta 1 Antibody (69310), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB839K
This item is not returnable.
View return policy
Description
IL-12 R beta 1 Monoclonal antibody specifically detects IL-12 R beta 1 in Human samples. It is validated for Flow Cytometry
Specifications
| IL-12 R beta 1 | |
| Monoclonal | |
| DyLight 488 | |
| CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL-12R subunit beta-1, IL12RB, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 1, Cys24-Glu540, Accession # P42701 | |
| 0.5 mL | |
| Cell Cycle and Replication | |
| 3594 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 69310 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction