Learn More
IL-12 R beta 2 Antibody (305719), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB19592L
Product Code. 30064348
  • € 607.00 / 0.10mL
Description
IL-12 R beta 2 Monoclonal antibody specifically detects IL-12 R beta 2 in Human, Mouse samples. It is validated for Direct ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-12 R beta 2
Monoclonal
DyLight 550
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse
Purified
Direct ELISA
305719
Direct ELISA
IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL-12R subunit beta-2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2
Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2, Cys28-Asn622, Accession # Q99665
0.1 mL
Cell Cycle and Replication
3595
Store at 4C in the dark.