missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13 Antibody (31606), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2131B
This item is not returnable.
View return policy
Description
IL-13 Monoclonal antibody specifically detects IL-13 in Human samples. It is validated for Western Blot, Neutralization, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| IL-13 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Neutralization, Flow Cytometry (Intracellular Staining), CyTOF | |
| 31606 | |
| Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
| E. coli-derived recombinant human IL-13, Gly35-Asn146, Accession # P35225 | |
| 0.1 mL | |
| Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3596 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction