Learn More
IL-13 Antibody (31606), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2131MFV500
Product Code. 18566026
  • ONLINE EXCLUSIVE PRICE
    € 708.00
    € 668.85 / 0.10mL
    Save € 39.15
    5% Off
Description
IL-13 Monoclonal antibody specifically detects IL-13 in Human samples. It is validated for Western Blot, Neutralization, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-13
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
0.1 mL
Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
3596
Store at 4C in the dark.
Western Blot, Neutralization, Flow Cytometry, CyTOF
31606
Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
E. coli-derived recombinant human IL-13, Gly35-Asn146, Accession # P35225