missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-13 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Product Code. 17835423
Change view
Click to view available options
Quantity:
2 x 500 μg
25 μg
50 μg
500 μg
Unit Size:
1mg
25µg
500µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17835423 25 μg 25µg
17804823 50 μg 50µg
17855593 2 x 500 μg 1mg
30054540 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17835423 Supplier PeproTech Supplier No. 500P178BT25UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-13. Anti-Murine IL-13-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mIL-13 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine IL-13 (500-P178) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mIL-13. Western Blot: To detect mIL-13 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mIL-13 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P178BT-1MG will be provided as 2 x 500 μg

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene Il13
Gene Accession No. P20109
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Gene Symbols Il13
Host Species Rabbit
Immunogen E.coli-derived Recombinant Murine IL-13.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16163
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.