missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13R alpha 1 Antibody (26), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89716PE
This item is not returnable.
View return policy
Description
IL-13R alpha 1 Monoclonal antibody specifically detects IL-13R alpha 1 in Human samples. It is validated for ELISA
Specifications
| IL-13R alpha 1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 26 | |
| ELISA | |
| bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-13R alpha 1 extracellular domain (Accession#: NP_001551.1; Met 1-Thr 343). | |
| 0.1 mL | |
| CD Markers, Immunology, Innate Immunity, Signal Transduction | |
| 3597 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction