missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13R alpha 2 Antibody (MM0367-11S23), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11663AF700
This item is not returnable.
View return policy
Description
IL-13R alpha 2 Monoclonal specifically detects IL-13R alpha 2 in Human samples. It is validated for Western Blot.
Specifications
| IL-13R alpha 2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IL13RA2 | |
| Human recombinant IL-13R alpha 2 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0367-11S23 | |
| Western Blot | |
| cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL-13R subunit alpha-2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein | |
| Mouse | |
| Protein G purified | |
| Cancer | |
| 3598 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction