Learn More
IL-13R alpha 2 Antibody (MM0367-11S23) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-11663-0.025MG
 View more versions of this product
Product Code. 18077576
  • € 206.00 / 25µg
Description
IL-13R alpha 2 Monoclonal specifically detects IL-13R alpha 2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-13R alpha 2
Monoclonal
LYOPH
Western Blot 1:500-1000
Q14627
IL13RA2
Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-13R a2 extracellular domain.
0.025 mg
Cancer
3598
Human
Purified
Western Blot
MM0367-11S23
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL-13R subunit alpha-2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein
Mouse
Protein G purified
RUO
Primary
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.