missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15 Antibody (111), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89449PE
This item is not returnable.
View return policy
Description
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for ELISA, ELISA
Specifications
| IL-15 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA | |
| 111 | |
| ELISA, Sandwich ELISA Detection | |
| IL-15MGC9721, interleukin 15, interleukin-15 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-15 (Accession#: NP_000576.1; Asn49-Ser162). | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity | |
| 3600 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction