Learn More
Novus Biologicals™ IL-15 Antibody (DISC0280) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28694-1mg
 View more versions of this product
Product Code. 30046407
  • € 920.00 / 1mg
Description
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-15
Monoclonal
Unconjugated
P40933
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
DISC0280
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
IL-15MGC9721, interleukin 15, interleukin-15
IL-15
1 mg
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
3600