missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Apolipoprotein CIII Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30043
This item is not returnable.
View return policy
Description
Apolipoprotein CIII Polyclonal antibody specifically detects Apolipoprotein CIII in Human samples. It is validated for ELISA,Western Blot
Specifications
| Apolipoprotein CIII | |
| Polyclonal | |
| PBS | |
| Goat | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| APOC3, apo-CIII, ApoC-III, APOCIII, Apolipoprotein C3, apolipoprotein C-III, MGC150353 | |
| Human Apolipoprotein CIII | |
| 1 mg | |
| Lipid and Metabolism | |
| 345 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction