missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15 Antibody (IL15/7048R) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14100JF585
This item is not returnable.
View return policy
Description
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IL-15 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| IL-15MGC9721, interleukin 15, interleukin-15 | |
| Recombinant fragment of human IL-15 (exact sequence is proprietary) (Uniprot: P40933) | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity | |
| 3600 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| IL15/7048R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction