missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15 Antibody (MM0368-13S34), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11665AF532
This item is not returnable.
View return policy
Description
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Western Blot, Neutralization
Specifications
| IL-15 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| IL-15MGC9721, interleukin 15, interleukin-15 | |
| Human recombinant IL-15 | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity | |
| 3600 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Neutralization | |
| MM0368-13S34 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction