missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15 Antibody (OTI2B4), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71034AF594
This item is not returnable.
View return policy
Description
IL-15 Monoclonal specifically detects IL-15 in Human samples. It is validated for Western Blot.
Specifications
| IL-15 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IL15 | |
| Recombinant protein fragment corresponding to amino acids 49-162 of human IL-15 (NP_751914) produced in E.coli. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI2B4 | |
| Western Blot | |
| IL-15MGC9721, interleukin 15, interleukin-15 | |
| Mouse | |
| 0.1 mL | |
| 3600 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction