missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15 Antibody (OTI2B4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71034FR
This item is not returnable.
View return policy
Description
IL-15 Monoclonal specifically detects IL-15 in Human samples. It is validated for Western Blot.
Specifications
| IL-15 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| IL15 | |
| Recombinant protein fragment corresponding to amino acids 49-162 of human IL-15 (NP_751914) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI2B4 | |
| Western Blot | |
| IL-15MGC9721, interleukin 15, interleukin-15 | |
| Mouse | |
| Affinity Purified | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity | |
| 3600.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction