missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15 Monoclonal Antibody (201136)

IL-15 Monoclonal Antibody (201136)
Brand: Invitrogen MA523786
This item is not returnable.
View return policy
Description
Specifications
| IL-15 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P48346 | |
| IL15 | |
| E. coli-derived recombinant mouse IL-15 Asn49-Ser162. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot | |
| 201136 | |
| Unconjugated | |
| IL15 | |
| AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 16168 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction