missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-15 Recombinant Rabbit Monoclonal Antibody (111), HRP, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 163.00 - € 510.00
Specifications
| Antigen | IL-15 |
|---|---|
| Clone | 111 |
| Concentration | 0.83 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019786
![]() |
Invitrogen™
MA552190 |
100 μg |
€ 510.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019571
|
Invitrogen™
MA552191 |
20 μg |
€ 163.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-15 Recombinant Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for ELISASpecifications
| IL-15 | |
| 0.83 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15 | |
| Recombinant Human IL-15/IL15/Interleukin 15 Protein (Asn49-Ser162). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| IL15 |
| 111 | |
| ELISA | |
| HRP | |
| Rabbit | |
| Human | |
| P40933 | |
| 3600 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title