Learn More
IL-15R alpha Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-41212CL1
Product Code. 30144944
  • € 965.00 / 0.10mL
Description
IL-15R alpha Polyclonal antibody specifically detects IL-15R alpha in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-15R alpha
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
ELISA, Western Blot
CoraFluor 1
CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179
Antibody was raised against a 16 amino acid peptide near the center of human IL-15RA. The immunogen is located within amino acids 40 - 90 of IL-15RA. Amino Acid Squence: NSGFKRKAGTSSLTE
0.1 mL
Cancer
3601
Store at 4°C in the dark. Do not freeze.