missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17/IL-17A Antibody (41802R), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3171RG
This item is not returnable.
View return policy
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| IL-17/IL-17A | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 41802R | |
| Western Blot, Intracellular Staining by Flow Cytometry | |
| CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| E.coli-derived recombinant human IL-17, Ile20-Ala155, Accession # Q16552 | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
| 3605 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction