Learn More
IL-17A Mouse anti-Human, Clone: OTI4A12, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA814599S
Encompass_Preferred
Product Code. 17569424
  • € 242.00 / 30µL
Description
IL-17A Monoclonal antibody specifically detects IL-17A in Human samples. It is validated for ELISA
Specifications
Specifications
IL-17A
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q16552
Mouse
Protein A/G
RUO
3605
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA
OTI4A12
Unconjugated
IL17A
Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A
Human recombinant protein fragment corresponding to amino acids 24-155 of human IL-17A (NP_002181) produced in E.coli.