Learn More
IL-17A Mouse, anti-Human, R718, Clone: N49-653, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566938
Description
Human IL-17A, also known as IL-17, is a proinflammatory cytokine that is encoded by the IL17A gene in chromosome 6. IL-17A is produced as a disulfide-linked homodimer comprised of two mature 136-amino acid polypeptides. It is a member of the IL-17 family of structurally related cytokines, designated IL-17A through IL-17F. Activated memory T cells, especially Th17 cells (specialized IL-17A-producing CD4+ T cells distinct from Th1 and Th2 cells) produce IL-17 and provide protective immunity against pathogens. Activated CD8+ T cells, γδT cells, NK cells and neutrophils can also be activated to produce IL-17A. IL-17A binds to and exerts its biological activity through IL-17 receptors (IL-17R) that are expressed by a variety of target cells including fibroblasts, epithelial and endothelial cells, monocytes/macrophages and mast cells. The ubiquitous IL-17R expression pattern may explain the broad tissue responsiveness to IL-17. IL-17 induces stromal cells to secrete cytokines and chemokines involved in inflammatory and hematopoietic processes. For example, IL-17 induces fibroblasts to produce IL-6, IL-8, G-CSF and express increased surface ICAM-1. The N49-653 antibody reacts with human IL-17A.
Specifications
| IL-17A | |
| Monoclonal | |
| R718 | |
| 100 Tests | |
| Primary | |
| Refrigerate Upon Arrival |
| Flow Cytometry | |
| N49-653 | |
| Mouse | |
| RUO | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.