missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17A Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | IL-17A |
|---|---|
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18005565
|
Proteintech
30754-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18014515
|
Proteintech
30754-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IL-17A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse IL-17A. This antibody recognizes mouse antigen. The IL-17A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IL-17A | |
| Western Blot, ELISA | |
| Rabbit | |
| 16171 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 500 μg/mL | |
| Liquid | |
| Mouse | |
| Ctla 8, Ctla8, Il17, Il17a, IL-17A, interleukin 17A | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title