Learn More
IL-17A Rat anti-Mouse, PE, Clone: TC11-18H10, BD
Rat Monoclonal Antibody
€ 149.92 - € 309.23
Specifications
| Antigen | IL-17A |
|---|---|
| Clone | TC11-18H10 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15880689
|
BD Biosciences
561020 |
25 μg |
€ 150.00 € 149.92 / Each Save € 0.08 0.05% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15879728
|
BD Biosciences
559502 |
0.1 mg |
€ 310.00 € 309.23 / Each Save € 0.77 0.25% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TC11-18H10 monoclonal antibody specifically binds to recombinant and natural mouse IL-17A proteins. IL-17A, also known as CTLA-8, is a T cell-derived cytokine that promotes inflammatory responses. Mouse IL-17A is a proinflammatory cytokine that can induce the release of IL-6 by mouse stromal cells. It has been shown to support the growth of hemopoietic progenitors in vitro; it can also stimulate granulopoiesis in vivo. The TC11-18H10 antibody has been reported to neutralize IL-17A activity. Recent studies have shown that IL-17A is produced by a unique subset of Th17 cells that develop along a pathway distinct from the Th1- and Th2- cell differentiation pathways. The mouse IL-17A cDNA was isolated from a cDNA library generated from TCR αβ +CD4-CD8- thymocytes.
Specifications
| IL-17A | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| IgG1 κ | |
| Affinity Purified |
| TC11-18H10 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.