missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-17A Recombinant Rabbit Monoclonal Antibody (217), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 109.00 - € 131.00
Specifications
| Antigen | IL-17A |
|---|---|
| Clone | 217 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019783
![]() |
Invitrogen™
MA552440 |
100 Tests |
€ 131.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019855
|
Invitrogen™
MA552441 |
25 Tests |
€ 109.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17A Recombinant Monoclonal antibody specifically detects IL-17A in Mouse samples. It is validated for Flow CytometrySpecifications
| IL-17A | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.03% ProClin 300 | |
| Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A | |
| Recombinant Mouse IL17 Protein (Thr22-Ala158). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| IL17A |
| 217 | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Mouse | |
| Q62386 | |
| 16171 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title