missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17B Antibody (B-B58), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18115AF532
This item is not returnable.
View return policy
Description
IL-17B Monoclonal antibody specifically detects IL-17B in Human samples. It is validated for ELISA
Specifications
| IL-17B | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20 | |
| Recombinant human IL-17B. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 27190 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| ELISA | |
| B-B58 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction