missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17B Antibody (MM0373-3D62), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11672APC
This item is not returnable.
View return policy
Description
IL-17B Monoclonal specifically detects IL-17B in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-17B | |
| Monoclonal | |
| APC | |
| Q9UHF5 | |
| IL17B | |
| Human recombinant IL-17B | |
| 0.1 mL | |
| Cytokine Research | |
| 27190 | |
| Human | |
| Purified |
| Flow Cytometry | |
| MM0373-3D62 | |
| Flow Cytometry | |
| Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects human IL-17B, but not IL-17A, C E or F. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction