missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17B Antibody (MM0373-3D62), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11672MFV610
This item is not returnable.
View return policy
Description
IL-17B Monoclonal antibody specifically detects IL-17B in Human samples. It is validated for Flow Cytometry
Specifications
| IL-17B | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| MM0373-3D62 | |
| Flow Cytometry | |
| Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20 | |
| Human recombinant IL-17B | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 27190 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction