Learn More
Novus Biologicals™ IL-17C Antibody (MOR106), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28300-1mg
 View more versions of this product
Product Code. 30048221
  • € 920.00 / 1mg
Description
IL-17C Monoclonal antibody specifically detects IL-17C in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-17C
Monoclonal
Unconjugated
Q9P0M4
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
MOR106
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CX2, Cytokine CX2, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401
IL-17c
1 mg
Cytokine Research
27189