missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17D Antibody (312724) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2274JF635
This item is not returnable.
View return policy
Description
IL-17D Monoclonal antibody specifically detects IL-17D in Mouse samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| IL-17D | |
| Monoclonal | |
| Janelia Fluor 635 | |
| FLJ30846, IL-17Dinterleukin 27, IL-22, IL-27interleukin-17D, IL27interleukin-27, interleukin 17D, Interleukin-27 | |
| E. coli-derived recombinant mouse IL-17D, Ala25-Arg205, Accession # NP_665836 | |
| 0.1 mL | |
| Cytokine Research | |
| 53342 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 312724 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction