missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.
Specifications
Specifications
| Antigen | IL-17D |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1D7H4 |
| Concentration | 1.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | IL17D |
| Gene Accession No. | Q8TAD2 |
| Gene Alias | IL 17D, IL 27, IL17D, IL27, interleukin 17D, Interleukin 27 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?