missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17E/IL-25 Antibody (182203) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1258JF585
This item is not returnable.
View return policy
Description
IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Human samples. It is validated for ELISA,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| IL-17E/IL-25 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
| E. coli-derived recombinant human IL-17E, Tyr33-Gly177, Accession # Q9H293 | |
| 0.1 mL | |
| Cytokine Research | |
| 64806 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 182203 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction