Learn More
IL-17E/IL-25 Antibody (8B1), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-27324DL594
Product Code. 18485948
  • € 516.00 / 0.10mL
Description
IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-17E/IL-25
Monoclonal
DyLight 594
IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25
Full-length human IL17E recombinant protein was used as the immunogen for this antibody.
0.1 mL
Cytokine Research
64806
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry
8B1
50mM Sodium Borate
Mouse
Protein G purified