missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17E/IL-25 Antibody (8B1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27324PCP
This item is not returnable.
View return policy
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-17E/IL-25 | |
| Monoclonal | |
| PerCP | |
| Q9H293 | |
| IL25 | |
| Full-length human IL17E recombinant protein was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Cytokine Research | |
| 64806 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| 8B1 | |
| Flow Cytometry 1 μL/1 million cells | |
| IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction