Learn More
Novus Biologicals™ IL-17E/IL-25 Antibody (Centocor patent anti-IL-25) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28161-50ug
 View more versions of this product
Product Code. 30044532
  • € 213.00 / 50µg
Description
IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-17E/IL-25
Monoclonal
Unconjugated
Q9H293
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Centocor patent anti-IL-25
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25
IL-25
50 μg
Cytokine Research
64806