missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17F Antibody (197301) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB13351JF635
This item is not returnable.
View return policy
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| IL-17F | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
| E. coli-derived recombinant human IL-17F, Arg31-Gln163, Accession # Q96PD4 | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 112744 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 197301 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction