missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17F Antibody (316016), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2057W
This item is not returnable.
View return policy
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| IL-17F | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 316016 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
| E. coli-derived recombinant mouse IL-17F, Arg29-Ala161, Accession # Q7TNI7 | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 112744 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction