missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17F Antibody (4H450), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27319IR
This item is not returnable.
View return policy
Description
IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for Western Blot.
Specifications
| IL-17F | |
| Monoclonal | |
| DyLight 755 | |
| Q96PD4 | |
| IL17F | |
| Full-length recombinant MRkIPkVGHT FFQkPESCPP VPGGSMkLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGkEDISMN SVPIQQETLV VRRkHQGCSV SFQLEkVLVT VGCTCVTPVI HHVQ | |
| 0.1 mL | |
| Cytokine Research | |
| 112744 | |
| Human | |
| Purified |
| Western Blot | |
| 4H450 | |
| Western Blot | |
| Cytokine ML-1, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Human. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction