missing translation for 'onlineSavingsMsg'
Learn More

IL-17F Antibody (B-F60), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-14589AF488

Product Code. 18594106

  • ONLINE EXCLUSIVE PRICE
    € 506.00
    € 477.75 / 0.10mL
    Save € 28.25
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-17F
Monoclonal
Alexa Fluor 488
Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F
Recombinant human IL-17F
0.1 mL
Biologically Active Proteins, Cytokine Research
112744
Store at 4°C in the dark.
IgG2b κ
ELISA
B-F60
50mM Sodium Borate
Mouse
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.