missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17F Antibody (B-F60), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14589MFV610
This item is not returnable.
View return policy
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for ELISA, Functional Assay, Enzyme-linked Immunosorbent Spot
Specifications
| IL-17F | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Functional Assay, ELISpot | |
| B-F60 | |
| ELISA, Functional, Enzyme-linked Immunosorbent Spot | |
| Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
| Recombinant human IL-17F | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 112744 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction