Learn More
IL-17RA/IL-17R Antibody (012), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89700H
Product Code. 18620061
  • € 660.00 / 0.10mL
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-17RA/IL-17R
Monoclonal
HRP
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA
012
ELISA
CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-17RA/IL-17R (NP_055154.3; Met 1-Trp 320).
0.1 mL
Innate Immunity
23765
Store at 4°C in the dark.